ENA295068576 Enamine

2-(3-ethylphenoxy)pyridine-4-carbonitrile

  • MDL number MFCD09938425
  • Cas Number : 1016887-17-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-64133

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-64133
Availability details for
Quantity
  • Availability
  • Add to Cart