ENA295068577 Enamine

N-(4-methylpentan-2-yl)-1H-indol-5-amine

  • MDL number MFCD11160658
  • Cas Number : 1042584-01-9
  • Molecular Weight : 216.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form oil
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-64134

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-64134
Availability details for
Quantity
  • Availability
  • Add to Cart