ENA295068589 Enamine

5-chloro-6-[(1-methyl-1H-imidazol-2-yl)sulfanyl]pyridine-3-carboxylic acid

  • MDL number MFCD11132219
  • Cas Number : 1019324-90-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-64161

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-64161
Availability details for
Quantity
  • Availability
  • Add to Cart