ENA295068590 Enamine

5-bromo-1-[2-(dimethylamino)ethyl]-2,3-dihydro-1H-indole-2,3-dione

  • MDL number MFCD11147950
  • Cas Number : 732962-63-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 102...104
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-64162

Safety Information

msds https://www.enaminestore.com/catalog/EN300-64162
Availability details for
Quantity
  • Availability
  • Add to Cart