ENA295068594 Enamine

4-benzylpiperidine-1-sulfonamide

  • MDL number MFCD11644030
  • Cas Number : 16168-15-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-64168

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-64168
Availability details for
Quantity
  • Availability
  • Add to Cart