ENA295068595 Enamine

azepane-1-sulfonamide

  • MDL number MFCD03078356
  • Cas Number : 4108-89-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-64169

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-64169
Availability details for
Quantity
  • Availability
  • Add to Cart