ENA295068597 Enamine

(dibutylsulfamoyl)amine

  • MDL number MFCD11205277
  • Cas Number : 53892-25-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-64171

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-64171
Availability details for
Quantity
  • Availability
  • Add to Cart