ENA295068610 Enamine

5-benzyl-2-hydroxybenzoic acid

  • MDL number MFCD08059184
  • Cas Number : 4386-41-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 139-140
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-61947

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-61947
Availability details for
Quantity
  • Availability
  • Add to Cart