ENA295068612 Enamine

4-bromo-2-nitrobenzene-1-sulfonyl chloride

  • MDL number MFCD12068352
  • Cas Number : 89465-98-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 95...97
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-39605

Safety Information

msds https://www.enaminestore.com/catalog/EN300-39605
Availability details for
Quantity
  • Availability
  • Add to Cart