ENA295068615 Enamine

1-cyclopropyl-5-methanesulfonyl-1H-1,2,3,4-tetrazole

  • MDL number MFCD11212377
  • Cas Number : 1094755-11-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-64191

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-64191
Availability details for
Quantity
  • Availability
  • Add to Cart