ENA295068622 Enamine

5-methyl-3-oxohexanenitrile

  • MDL number MFCD01960834
  • Cas Number : 64373-43-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Product Link https://www.enaminestore.com/catalog/EN300-64199

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-64199
Availability details for
Quantity
  • Availability
  • Add to Cart