ENA295068626 Enamine

2-amino-2-propylpentanenitrile

  • MDL number MFCD09813845
  • Cas Number : 805181-81-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-64203

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-64203
Availability details for
Quantity
  • Availability
  • Add to Cart