ENA295068632 Enamine

3,4-dimethyl-6H,7H-[1,2]oxazolo[3,4-d]pyridazin-7-one

  • MDL number MFCD00126308
  • Cas Number : 15911-16-7
  • Molecular Weight : 165.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-62102

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-62102
Availability details for
Quantity
  • Availability
  • Add to Cart