ENA295068635 Enamine

2-methyl-4-[(5-methyl-1,3,4-thiadiazol-2-yl)sulfanyl]aniline

  • MDL number MFCD12800069
  • Cas Number : 1178072-62-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-64229

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-64229
Availability details for
Quantity
  • Availability
  • Add to Cart