ENA295068637 Enamine

1-[2-(4-methoxyphenyl)ethyl]imidazolidine-2,4,5-trione

  • MDL number MFCD11544035
  • Cas Number : 1094408-14-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-64232

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-64232
Availability details for
Quantity
  • Availability
  • Add to Cart