ENA295068643 Enamine

2-chloro-4-(1H-imidazol-2-yl)pyridine

  • MDL number MFCD11501094
  • Cas Number : 1006589-03-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-64240

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-64240
Availability details for
Quantity
  • Availability
  • Add to Cart