ENA295068647 Enamine

1-(1-methyl-2-oxo-1,2-dihydropyridine-4-carbonyl)piperidine-3-carboxylic acid

  • MDL number MFCD12656807
  • Cas Number : 1099016-99-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-64245

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-64245
Availability details for
Quantity
  • Availability
  • Add to Cart