ENA295068650 Enamine

ethyl 1-(1-methyl-2-oxo-1,2-dihydropyridine-4-carbonyl)piperidine-3-carboxylate

  • MDL number MFCD13690840
  • Cas Number : 1153394-19-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-64251

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-64251
Availability details for
Quantity
  • Availability
  • Add to Cart