ENA295068653 Enamine

1-(4-bromophenyl)-4,4-dimethylpentan-3-one

  • MDL number MFCD11655203
  • Cas Number : 1099621-38-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-64254

Safety Information

msds https://www.enaminestore.com/catalog/EN300-64254
Availability details for
Quantity
  • Availability
  • Add to Cart