ENA295068658 Enamine

1-(2-bromophenyl)-4,4-dimethylpentan-3-one

  • MDL number MFCD11655217
  • Cas Number : 252935-80-1
  • Molecular Weight : 269.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-64259

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-64259
Availability details for
Quantity
  • Availability
  • Add to Cart