ENA295068659 Enamine

7-(methylsulfanyl)-2,3-dihydro-1,4-benzodioxin-6-amine

  • MDL number MFCD12823585
  • Cas Number : 1183512-25-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-64260

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-64260
Availability details for
Quantity
  • Availability
  • Add to Cart