ENA295068664 Enamine

3-methylfuran-2-carboxamide

  • MDL number MFCD11643939
  • Cas Number : 84374-70-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-64267

Safety Information

msds https://www.enaminestore.com/catalog/EN300-64267
Availability details for
Quantity
  • Availability
  • Add to Cart