ENA295068665 Enamine

N-cyclopentyl-4-nitroaniline

  • MDL number MFCD01463611
  • Cas Number : 342044-45-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 86...88
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-64268

Safety Information

msds https://www.enaminestore.com/catalog/EN300-64268
Availability details for
Quantity
  • Availability
  • Add to Cart