ENA295068671 Enamine

methyl 2-[2-(2,6-dichlorophenyl)-N-methylacetamido]acetate

  • MDL number MFCD12643619
  • Cas Number : 1153115-13-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-64300

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-64300
Availability details for
Quantity
  • Availability
  • Add to Cart