ENA295068677 Enamine

1-phenyl-N-propylpiperidin-4-amine

  • MDL number MFCD11640903
  • Cas Number : 1096836-52-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-64309

Safety Information

msds https://www.enaminestore.com/catalog/EN300-64309
Availability details for
Quantity
  • Availability
  • Add to Cart