ENA295068681 Enamine

5-[chloro(phenyl)methyl]-3-methyl-1,2,4-oxadiazole

  • MDL number MFCD11101520
  • Cas Number : 1152582-45-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-64317

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-64317
Availability details for
Quantity
  • Availability
  • Add to Cart