ENA295068684 Enamine

3-[1-(ethanesulfonyl)piperidin-4-yl]-1-(pyridin-3-ylmethyl)urea

  • MDL number MFCD16547658
  • Cas Number : 1240526-69-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95

Safety Information

msds https://www.enaminestore.com/catalog/EN300-64329
Availability details for
Quantity
  • Availability
  • Add to Cart