ENA295068687 Enamine

1-methyl-1,2,3,4-tetrahydronaphthalene-1-carboxylic acid

  • MDL number MFCD16547661
  • Cas Number : 26516-28-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 114-119
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-64333

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-64333
Availability details for
Quantity
  • Availability
  • Add to Cart