ENA295068689 Enamine

2-methyl-4-[(4-methyl-1,3-thiazol-2-yl)sulfanyl]aniline

  • MDL number MFCD12169276
  • Cas Number : 1155909-82-2
  • Molecular Weight : 236.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-64335

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-64335
Availability details for
Quantity
  • Availability
  • Add to Cart