ENA295068692 Enamine

4-(3-methoxyphenyl)-1H-pyrrole-3-carboxylic acid

  • MDL number MFCD11217947
  • Cas Number : 1096790-29-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-64338

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-64338
Availability details for
Quantity
  • Availability
  • Add to Cart