ENA295068695 Enamine

2-amino-4,5,6,7-tetrahydro-1,3-benzothiazol-7-one

  • MDL number MFCD00460519
  • Cas Number : 17583-10-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 265-268
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-64342

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-64342
Availability details for
Quantity
  • Availability
  • Add to Cart