ENA295068704 Enamine

thiophene-2,4-dicarboxylic acid

  • MDL number MFCD00159563
  • Cas Number : 36157-39-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 279...281
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-64354

Safety Information

msds https://www.enaminestore.com/catalog/EN300-64354
Availability details for
Quantity
  • Availability
  • Add to Cart