ENA295068706 Enamine

4-{[(1-cyclopropylethyl)amino]methyl}phenol

  • MDL number MFCD12193509
  • Cas Number : 1155988-12-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-64358

Safety Information

msds https://www.enaminestore.com/catalog/EN300-64358
Availability details for
Quantity
  • Availability
  • Add to Cart