ENA295068709 Enamine

2-(4-bromo-1H-pyrazol-1-yl)-2-methylpropanoic acid

  • MDL number MFCD04967795
  • Cas Number : 917569-72-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-62246

Safety Information

msds https://www.enaminestore.com/catalog/EN300-62246
Availability details for
Quantity
  • Availability
  • Add to Cart