ENA295068713 Enamine

7-methoxy-3-methyl-1-benzofuran-2-carboxylic acid

  • MDL number MFCD11205101
  • Cas Number : 18703-80-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-64400

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-64400
Availability details for
Quantity
  • Availability
  • Add to Cart