ENA295068721 Enamine

2-(2-cyclopentylacetamido)acetic acid

  • MDL number MFCD09043376
  • Cas Number : 926217-69-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-62268

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-62268
Availability details for
Quantity
  • Availability
  • Add to Cart