ENA295068729 Enamine

6-bromo-8-fluoro-4-hydroxyquinoline-3-carboxylic acid

  • MDL number MFCD09861321
  • Cas Number : 1019016-07-9
  • Molecular Weight : 286.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-64420

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-64420
Availability details for
Quantity
  • Availability
  • Add to Cart