ENA295068732 Enamine

methyl 2-[2-(2-chloro-6-fluorophenyl)-N-methylacetamido]acetate

  • MDL number MFCD12643622
  • Cas Number : 1156981-49-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form oil
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-64423

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-64423
Availability details for
Quantity
  • Availability
  • Add to Cart