ENA295068733 Enamine

[(3-fluorophenyl)(1-methyl-1H-pyrazol-4-yl)methyl](methyl)amine

  • MDL number MFCD12182634
  • Cas Number : 1153244-33-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form oil
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-64424

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-64424
Availability details for
Quantity
  • Availability
  • Add to Cart