ENA295068736 Enamine

6-[ethyl(propyl)amino]pyridine-3-carboxylic acid

  • MDL number MFCD11626108
  • Cas Number : 1094866-00-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-62304

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-62304
Availability details for
Quantity
  • Availability
  • Add to Cart