ENA295068740 Enamine

4-(tert-butylsulfanyl)-2-methylaniline

  • MDL number MFCD12169280
  • Cas Number : 75794-23-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-64442

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-64442
Availability details for
Quantity
  • Availability
  • Add to Cart