ENA295068748 Enamine

[4-(propan-2-yl)phenyl]methanethiol

  • MDL number MFCD11127065
  • Cas Number : 4427-53-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Product Link https://www.enaminestore.com/catalog/EN300-64452

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-64452
Availability details for
Quantity
  • Availability
  • Add to Cart