ENA295068752 Enamine

N-ethyl-2-(quinazolin-4-ylamino)acetamide

  • MDL number MFCD13233853
  • Cas Number : 1179600-29-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-64456

Safety Information

msds https://www.enaminestore.com/catalog/EN300-64456
Availability details for
Quantity
  • Availability
  • Add to Cart