ENA295068754 Enamine

1-N-(oxan-4-yl)benzene-1,4-diamine

  • MDL number MFCD12088410
  • Cas Number : 1153226-71-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-64461

Safety Information

msds https://www.enaminestore.com/catalog/EN300-64461
Availability details for
Quantity
  • Availability
  • Add to Cart