ENA295068756 Enamine

N-(4-aminophenyl)-3-(piperidin-1-yl)propanamide

  • MDL number MFCD09729342
  • Cas Number : 738546-03-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-62314

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-62314
Availability details for
Quantity
  • Availability
  • Add to Cart