ENA295068770 Enamine

5-ethyl-6-hydroxy-3-(4-methoxyphenyl)-2-sulfanyl-3,4-dihydropyrimidin-4-one

  • MDL number MFCD16547545
  • Cas Number : 1240526-41-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-62365

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-62365
Availability details for
Quantity
  • Availability
  • Add to Cart