ENA295068773 Enamine

3-[(thiophen-2-ylmethyl)sulfamoyl]thiophene-2-carboxylic acid

  • MDL number MFCD09455215
  • Cas Number : 944894-47-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 138...140
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-62372

Safety Information

msds https://www.enaminestore.com/catalog/EN300-62372
Availability details for
Quantity
  • Availability
  • Add to Cart