ENA295068779 Enamine

3-{[(9-ethyl-9H-carbazol-3-yl)methyl]amino}propan-1-ol

  • MDL number MFCD13684245
  • Cas Number : 1157070-06-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-64498

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-64498
Availability details for
Quantity
  • Availability
  • Add to Cart