ENA295068781 Enamine

2-amino-5-chloro-N-(pyridin-4-yl)benzamide

  • MDL number MFCD09042865
  • Cas Number : 926194-69-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-62399

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-62399
Availability details for
Quantity
  • Availability
  • Add to Cart