ENA295068785 Enamine

4-(3-fluorobenzoyl)-1-methyl-1H-pyrazole

  • MDL number MFCD11211326
  • Cas Number : 1152836-01-5
  • Molecular Weight : 204.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-62421

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-62421
Availability details for
Quantity
  • Availability
  • Add to Cart